Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-3-(naphthalen-1-yl)propanoic acid | 28095-56-9 | MFCD00067507 | 98.5% | 215.25 | C13H13NO2 | 5 G
SDP

Catalog No. 5000301197
Encompass_Preferred

2-Amino-3-(naphthalen-1-yl)propanoic acid is an alanine-derived amino acid analogue supplied as a research reagent. It is used as a substituted amino acid probe in biochemical and pharmacological studies. The compound has documented molecular formula C13H13NO2 and molecular weight 215.25, is provided as a powder with specified purity, and includes datasheet and safety documentation for handling.

  • Alanine-derived amino acid analogue for research applications.
  • Molecular formula C13H13NO2, molecular weight 215.25.
  • Purity 98.47%.
  • Available in multiple package sizes including 5 g, 10 g, 25 g, 100 g.
  • Powder storage: -20°C (up to 3 years) and 4°C (up to 2 years).
  • In solvent storage: -80°C (up to 6 months) and -20°C (up to 1 month).
  • Data sheet and SDS available for handling and safety information.

Catalog No. 50-003-01197 Supplier Medchemexpress LLC Supplier No. HYW0397635G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-3-(naphthalen-1-yl)propanoic acid is an alanine-derived amino acid analogue supplied as a research reagent. It is used as a substituted amino acid probe in biochemical and pharmacological studies. The compound has documented molecular formula C13H13NO2 and molecular weight 215.25, is provided as a powder with specified purity, and includes datasheet and safety documentation for handling.

  • Alanine-derived amino acid analogue for research applications.
  • Molecular formula C13H13NO2, molecular weight 215.25.
  • Purity 98.47%.
  • Available in multiple package sizes including 5 g, 10 g, 25 g, 100 g.
  • Powder storage: -20°C (up to 3 years) and 4°C (up to 2 years).
  • In solvent storage: -80°C (up to 6 months) and -20°C (up to 1 month).
  • Data sheet and SDS available for handling and safety information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.