Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-3-(naphthalen-1-yl)propanoic acid | 28095-56-9 | MFCD00067507 | 98.5% | 215.25 g/mol | C13H13NO2 | 10 G
SDP

Catalog No. 5000299479
Encompass_Preferred

2-Amino-3-(naphthalen-1-yl)propanoic acid is an alanine derivative featuring a naphthyl substituent at the 3-position of the propanoic-acid backbone. It is intended for research use as an amino-acid building block and reagent in synthetic and biological studies.

  • High purity (98.47%).
  • Molecular weight 215.25 g/mol.
  • Chemical formula C13H13NO2.
  • Available in multiple package sizes, including 10 g.
  • Certificate of analysis and data sheet available.
  • Intended for research use only, not for human use.

Catalog No. 50-002-99479 Supplier Medchemexpress LLC Supplier No. HYW03976310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-3-(naphthalen-1-yl)propanoic acid is an alanine derivative featuring a naphthyl substituent at the 3-position of the propanoic-acid backbone. It is intended for research use as an amino-acid building block and reagent in synthetic and biological studies.

  • High purity (98.47%).
  • Molecular weight 215.25 g/mol.
  • Chemical formula C13H13NO2.
  • Available in multiple package sizes, including 10 g.
  • Certificate of analysis and data sheet available.
  • Intended for research use only, not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.