Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-3-(naphthalen-1-yl)propanoic acid | 28095-56-9 | MFCD00067507 | 98.5% | 215.25 g/mol | C13H13NO2 | 100 G
SDP

Catalog No. 5000300537
Encompass_Preferred

2-amino-3-(naphthalen-1-yl)propanoic acid is an alanine derivative bearing a naphthyl substituent on the beta carbon. It is supplied for research use and is used as an amino acid derivative and building block in biochemical and pharmacological studies. Handle with standard laboratory precautions and consult safety data before use.

  • CAS number 28095-56-9.
  • Molecular formula C13H13NO2.
  • Molecular weight 215.25 g/mol.
  • Purity 98.5%.
  • Suitable as a building block in peptide and small-molecule synthesis.

Catalog No. 50-003-00537 Supplier Medchemexpress LLC Supplier No. HYW039763100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-amino-3-(naphthalen-1-yl)propanoic acid is an alanine derivative bearing a naphthyl substituent on the beta carbon. It is supplied for research use and is used as an amino acid derivative and building block in biochemical and pharmacological studies. Handle with standard laboratory precautions and consult safety data before use.

  • CAS number 28095-56-9.
  • Molecular formula C13H13NO2.
  • Molecular weight 215.25 g/mol.
  • Purity 98.5%.
  • Suitable as a building block in peptide and small-molecule synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.