Learn More
Medchemexpress LLC 2-amino-3-(naphthalen-1-yl)propanoic acid | 28095-56-9 | MFCD00067507 | 98.5% | 215.25 g/mol | C13H13NO2 | 25 G
SDP
Catalog No. 5000302902
Encompass_Preferred
2-Amino-3-(naphthalen-1-yl)propanoic acid is an alanine-derived amino acid analogue bearing a naphthyl substituent at the 3-position. Supplied for research use, it is commonly used as a synthetic building block and reference compound in organic synthesis, medicinal chemistry, and structure-activity relationship studies.
  • Alanine-derived amino acid analogue bearing a naphthyl substituent.
  • Listed purity of 98.47% suitable for research applications.
  • Molecular formula C13H13NO2 and molecular weight 215.25 g/mol.
  • Used as a synthetic building block and reference in medicinal chemistry and SAR studies.
  • Provided for research use only; SDS and product data sheet available.
  • Available in multi-gram package sizes for laboratory use.
Catalog No. 50-003-02902 Supplier Medchemexpress LLC Supplier No. 25GHYW039763
2-Amino-3-(naphthalen-1-yl)propanoic acid is an alanine-derived amino acid analogue bearing a naphthyl substituent at the 3-position. Supplied for research use, it is commonly used as a synthetic building block and reference compound in organic synthesis, medicinal chemistry, and structure-activity relationship studies.
  • Alanine-derived amino acid analogue bearing a naphthyl substituent.
  • Listed purity of 98.47% suitable for research applications.
  • Molecular formula C13H13NO2 and molecular weight 215.25 g/mol.
  • Used as a synthetic building block and reference in medicinal chemistry and SAR studies.
  • Provided for research use only; SDS and product data sheet available.
  • Available in multi-gram package sizes for laboratory use.