Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-pyridinemethanol 96.0+%, TCI America™
SDP

Catalog No. A23585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23585G 5 g
A23581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23585G Supplier TCI America Supplier No. A23585G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 23612-57-9
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.14
MDL Number MFCD05663510
InChI Key FEIACFYXEWBKHU-UHFFFAOYSA-N
Synonym 2-aminopyridin-3-yl methanol, 2-aminopyridine-3-methanol, 2-amino-3-hydroxymethylpyridine, 2-amino-pyridin-3-yl-methanol, 2-amino-3-pyridinyl methanol, 2-amino-3-pyridinemethanol, 2-amino-3-hydroxymethyl pyridine, 3-pyridinemethanol, 2-amino, 2-aminopyridine-3-yl-methanol
PubChem CID 11159325
IUPAC Name (2-aminopyridin-3-yl)methanol
SMILES NC1=NC=CC=C1CO

Specifications

Melting Point 67°C
Color Yellow
Boiling Point 279°C
Quantity 5 g
Formula Weight 124.14
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-pyridinemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.