Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-(trifluoromethyl)pyridine, 97%

Catalog No. AAH3430314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3430314 25 g
AAH3430306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3430314 Supplier Thermo Scientific Chemicals Supplier No. H3430314
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 183610-70-0 | C6H5F3N2 | 162.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 183610-70-0
Molecular Formula C6H5F3N2
Molecular Weight (g/mol) 162.12
MDL Number MFCD04972538
InChI Key YWOWJQMFMXHLQD-UHFFFAOYSA-N
Synonym 2-amino-3-trifluoromethyl pyridine, 3-trifluoromethyl pyridin-2-amine, 3-trifluoromethyl-2-pyridinamine, 2-amino-3-trifluoromethylpyridine, 3-trifluoromethyl-pyridin-2-ylamine, 2-pyridinamine, 3-trifluoromethyl, 3-trifluoromethyl-2-pyridylamine, pubchem9819, acmc-1c6sx, ksc495e5p
PubChem CID 2760920
IUPAC Name 3-(trifluoromethyl)pyridin-2-amine
SMILES NC1=C(C=CC=N1)C(F)(F)F

Specifications

Melting Point 70°C to 72°C
Quantity 25 g
UN Number UN2811
Formula Weight 162.11
Percent Purity 97%
Chemical Name or Material 2-Amino-3-(trifluoromethyl)pyridine
Hazard Category H301-H312-H315-H319-H335
Hazard Statement GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.