Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-(4-chlorophenyl)thiazole, 98%

Catalog No. AAL1406706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1406706 5 g
AAL1406703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1406706 Supplier Thermo Scientific Chemicals Supplier No. L1406706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2103-99-3 | C9H7ClN2S | 210.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2103-99-3
Molecular Formula C9H7ClN2S
Molecular Weight (g/mol) 210.68
MDL Number MFCD00047058
InChI Key DWGWNNCHJPKZNC-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl thiazol-2-amine, 4-4-chlorophenyl-1,3-thiazol-2-amine, 2-amino-4-4-chlorophenyl thiazole, 2-thiazolamine, 4-4-chlorophenyl, 4-4-chloro-phenyl-thiazol-2-ylamine, 4-4-chlorophenyl-2-thiazolamine, 2-amino 4 p-chloro phenyl thiazole, 4-4-chlorophenyl-1,3-thiazole-2-ylamine, p-chloro phenyl-4 amino-2 thiazole french, pubchem9961
PubChem CID 73241
IUPAC Name 4-(4-chlorophenyl)-1,3-thiazol-2-amine
SMILES NC1=NC(=CS1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 168°C to 170°C
Quantity 5 g
Beilstein 153889
Formula Weight 210.69
Percent Purity 98%
Chemical Name or Material 2-Amino-4-(4-chlorophenyl)thiazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber XJ1026732
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.