Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-4-(4-pyridyl)-thiazole | 30235-28-0 | 98.9% | 177.23 | C8H7N3S | 1 G
SDP

Catalog No. 5000210204
Encompass_Preferred

2-amino-4-(4-pyridyl)-thiazole is a synthetic intermediate used in pharmaceutical and medicinal chemistry research. It is supplied for research use; purity, batch, and storage details are provided on the certificate of analysis.

  • Synthetic intermediate for medicinal chemistry and pharmaceutical synthesis.
  • Molecular formula: C8H7N3S.
  • Molecular weight: 177.23 g/mol.
  • High purity reported by HPLC (98.9%).
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-10204 Supplier Medchemexpress LLC Supplier No. HYW0448051G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-amino-4-(4-pyridyl)-thiazole is a synthetic intermediate used in pharmaceutical and medicinal chemistry research. It is supplied for research use; purity, batch, and storage details are provided on the certificate of analysis.

  • Synthetic intermediate for medicinal chemistry and pharmaceutical synthesis.
  • Molecular formula: C8H7N3S.
  • Molecular weight: 177.23 g/mol.
  • High purity reported by HPLC (98.9%).
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.