Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-4-(4-pyridyl)-thiazole | 30235-28-0 | MFCD00490009 | 98.9% | 177.23 g/mol | C8H7N3S | 500 MG
SDP

Catalog No. 5000210625
Encompass_Preferred

2-amino-4-(4-pyridyl)-thiazole is a small-molecule synthetic intermediate used in pharmaceutical and medicinal chemistry synthesis. The compound is supplied as a characterized solid with supporting batch COA data.

  • High purity: 98.9% (HPLC).
  • Physical form: off-white to pink solid.
  • Molecular formula: C8H7N3S.
  • Molecular weight: 177.23 g/mol.
  • Intended use: synthetic intermediate for medicinal chemistry and pharmaceutical synthesis.

Catalog No. 50-002-10625 Supplier Medchemexpress LLC Supplier No. HYW044805500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-amino-4-(4-pyridyl)-thiazole is a small-molecule synthetic intermediate used in pharmaceutical and medicinal chemistry synthesis. The compound is supplied as a characterized solid with supporting batch COA data.

  • High purity: 98.9% (HPLC).
  • Physical form: off-white to pink solid.
  • Molecular formula: C8H7N3S.
  • Molecular weight: 177.23 g/mol.
  • Intended use: synthetic intermediate for medicinal chemistry and pharmaceutical synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.