Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Amino-4',5'-dimethoxyacetophenone 98.0+%, TCI America™
SDP

Catalog No. A28425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28425G 5 g
A28421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28425G Supplier TCI America Supplier No. A28425G

Chemical Identifiers

CAS 4101-30-8
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD00016646
InChI Key KGKWXEGYKGTMAK-UHFFFAOYSA-N
Synonym 1-2-amino-4,5-dimethoxyphenyl ethanone, 2'-amino-4',5'-dimethoxyacetophenone, 2-amino-4,5-dimethoxyacetophenone, 1-2-amino-4,5-dimethoxyphenyl ethan-1-one, 1-2-amino-4,5-dimethoxyhenyl ethanone, ethanone, 1-2-amino-4,5-dimethoxyphenyl, 1-acetyl-2-amino-4,5-dimethoxybenzene, pubchem4007, acmc-1arib, 2-acetyl-4,5-dimethoxyaniline
PubChem CID 602085
IUPAC Name 1-(2-amino-4,5-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1N)OC)OC

Specifications

Melting Point 107°C
Color Yellow
Quantity 5 g
Formula Weight 195.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Amino-4′,5′-dimethoxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.