Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4,5-dimethylthiazole Hydrobromide 98.0+%, TCI America™
SDP

Catalog No. A20625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20625G 5 g
A206225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20625G Supplier TCI America Supplier No. A20625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7170-76-5
Molecular Formula C5H9BrN2S
Molecular Weight (g/mol) 209.105
MDL Number MFCD00035212
InChI Key CATCJRCKBGAMKK-UHFFFAOYSA-N
Synonym 2-amino-4,5-dimethylthiazole hydrobromide, 4,5-dimethyl-1,3-thiazol-2-amine hydrobromide, 4,5-dimethylthiazol-2-amine hydrobromide, unii-nm8hvb696p, 2-thiazolamine, 4,5-dimethyl-, monohydrobromide, nm8hvb696p, 2-thiazolamine, 4,5-dimethyl-, hydrobromide 1:1, dimethyl-1,3-thiazol-2-amine hydrobromide, thiazole, 2-amino-4,5-dimethyl-, hydrobromide, acmc-209ojv
PubChem CID 73237
IUPAC Name 4,5-dimethyl-1,3-thiazol-2-amine;hydrobromide
SMILES CC1=C(SC(=N1)N)C.Br

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 209.11
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4,5-dimethylthiazole Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.