Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Amino-4-bromobenzoic acid | 20776-50-5 | 216.03 | 1 ML
SDP

Catalog No. 5000448692
Encompass_Preferred

2-Amino-4-bromobenzoic acid is a biochemical reagent used as a biological material or organic compound for life science research. It serves as an important raw material and intermediate in organic synthesis, pharmaceuticals, agrochemicals, and dyestuff.

  • Used as a biochemical reagent
  • Important raw material and intermediate
  • Suitable for organic synthesis, pharmaceuticals, agrochemicals, and dyestuff
  • Purity of 99.95%
  • Molecular weight of 216.03
  • Available in solution form
  • White to yellow solid appearance

Catalog No. 50-004-48692 Supplier Medchemexpress LLC Supplier No. HYW00309310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-4-bromobenzoic acid is a biochemical reagent used as a biological material or organic compound for life science research. It serves as an important raw material and intermediate in organic synthesis, pharmaceuticals, agrochemicals, and dyestuff.

  • Used as a biochemical reagent
  • Important raw material and intermediate
  • Suitable for organic synthesis, pharmaceuticals, agrochemicals, and dyestuff
  • Purity of 99.95%
  • Molecular weight of 216.03
  • Available in solution form
  • White to yellow solid appearance

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.