Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Amino-4-bromobenzoic acid | 20776-50-5 | 216.03 | 1000 G
SDP

Catalog No. 5000445633
Encompass_Preferred

2-Amino-4-bromobenzoic acid is a high-purity biochemical reagent. It serves as a biological material or organic compound for various life science research applications and is a key intermediate in organic synthesis, pharmaceuticals, agrochemicals, and dyestuff production.

  • Purity: 99.95%
  • White to yellow solid appearance
  • Chemical formula: C7H6BrNO2
  • Molecular weight: 216.03
  • CAS No.: 20776-50-5
  • Suitable for life science research
  • Used in organic synthesis, pharmaceuticals, agrochemicals, and dyestuff
  • Store at 4°C, protected from light

Catalog No. 50-004-45633 Supplier Medchemexpress LLC Supplier No. HYW0030931000G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-4-bromobenzoic acid is a high-purity biochemical reagent. It serves as a biological material or organic compound for various life science research applications and is a key intermediate in organic synthesis, pharmaceuticals, agrochemicals, and dyestuff production.

  • Purity: 99.95%
  • White to yellow solid appearance
  • Chemical formula: C7H6BrNO2
  • Molecular weight: 216.03
  • CAS No.: 20776-50-5
  • Suitable for life science research
  • Used in organic synthesis, pharmaceuticals, agrochemicals, and dyestuff
  • Store at 4°C, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.