Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Amino-4-bromobenzoic acid | 20776-50-5 | 216.03 | 5 G
SDP

Catalog No. 5000447453
Encompass_Preferred

2-Amino-4-bromobenzoic acid is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is used as an important raw material and intermediate in organic synthesis, pharmaceuticals, agrochemicals, and dyestuffs.

  • High purity: 99.95%
  • Molecular weight: 216.03
  • CAS number: 20776-50-5
  • Appearance: Solid
  • Color: White to yellow
  • Suitable for organic synthesis, pharmaceuticals, agrochemicals, and dyestuffs

Catalog No. 50-004-47453 Supplier Medchemexpress LLC Supplier No. HYW0030935G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-4-bromobenzoic acid is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is used as an important raw material and intermediate in organic synthesis, pharmaceuticals, agrochemicals, and dyestuffs.

  • High purity: 99.95%
  • Molecular weight: 216.03
  • CAS number: 20776-50-5
  • Appearance: Solid
  • Color: White to yellow
  • Suitable for organic synthesis, pharmaceuticals, agrochemicals, and dyestuffs

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.