Learn More
Medchemexpress LLC 2-Amino-4-bromobenzoic acid | 20776-50-5 | 216.03 | 500 G
SDP
Catalog No. 5000448641
Encompass_Preferred
2-Amino-4-bromobenzoic acid is a biochemical reagent primarily used in life science research. It functions as a biological material or organic compound and serves as a crucial raw material and intermediate in various organic syntheses, including pharmaceuticals, agrochemicals, and dyestuff production. This product is intended for research use only.
  • Biochemical reagent for life science research
  • Important raw material in organic synthesis
  • Intermediate in pharmaceuticals, agrochemicals, and dyestuff production
  • High purity of 99.9%
Catalog No. 50-004-48641 Supplier Medchemexpress LLC Supplier No. HYW003093500G
2-Amino-4-bromobenzoic acid is a biochemical reagent primarily used in life science research. It functions as a biological material or organic compound and serves as a crucial raw material and intermediate in various organic syntheses, including pharmaceuticals, agrochemicals, and dyestuff production. This product is intended for research use only.
  • Biochemical reagent for life science research
  • Important raw material in organic synthesis
  • Intermediate in pharmaceuticals, agrochemicals, and dyestuff production
  • High purity of 99.9%