Learn More
Medchemexpress LLC 2-Amino-4-bromopyridine | 84249-14-9 | 100.0% | 173.01 | 50 G
SDP
Catalog No. 5000403844
Encompass_Preferred
2-Amino-4-bromopyridine is an important intermediate used for the synthesis of other active compounds. Its derivative compounds can inhibit tumor cell proliferation by inducing G1 phase arrest and effectively act on wild-type ALK and drug-resistant mutants.
  • Important intermediate for synthesis
  • Derivatives can inhibit tumor cell proliferation
  • Induces G1 phase arrest
  • Effectively acts on wild-type ALK and drug-resistant mutants
Catalog No. 50-004-03844 Supplier Medchemexpress LLC Supplier No. HY4100050G
2-Amino-4-bromopyridine is an important intermediate used for the synthesis of other active compounds. Its derivative compounds can inhibit tumor cell proliferation by inducing G1 phase arrest and effectively act on wild-type ALK and drug-resistant mutants.
  • Important intermediate for synthesis
  • Derivatives can inhibit tumor cell proliferation
  • Induces G1 phase arrest
  • Effectively acts on wild-type ALK and drug-resistant mutants