Learn More
Medchemexpress LLC 2-Amino-4-bromopyridine | 84249-14-9 | 99.9% | 173.01 | 100 G
SDP
Catalog No. 5000405158
Encompass_Preferred
2-Amino-4-bromopyridine is an important intermediate chemical compound utilized in the synthesis of other active compounds. It can be specifically used for the synthesis of 2-aminopyridine derivatives. These derivatives have shown potential to inhibit tumor cell proliferation by inducing G1 phase arrest and are effective against both wild-type ALK and drug-resistant mutants. The product appears as an off-white to yellow solid.
  • Important intermediate for synthesis of active compounds
  • Used for synthesizing 2-aminopyridine derivatives
  • Derivatives inhibit tumor cell proliferation
  • Derivatives induce G1 phase arrest
  • Effective against wild-type ALK and drug-resistant mutants
Catalog No. 50-004-05158 Supplier Medchemexpress LLC Supplier No. HY41000100G
2-Amino-4-bromopyridine is an important intermediate chemical compound utilized in the synthesis of other active compounds. It can be specifically used for the synthesis of 2-aminopyridine derivatives. These derivatives have shown potential to inhibit tumor cell proliferation by inducing G1 phase arrest and are effective against both wild-type ALK and drug-resistant mutants. The product appears as an off-white to yellow solid.
  • Important intermediate for synthesis of active compounds
  • Used for synthesizing 2-aminopyridine derivatives
  • Derivatives inhibit tumor cell proliferation
  • Derivatives induce G1 phase arrest
  • Effective against wild-type ALK and drug-resistant mutants