Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-chloro-5-nitrophenol 98.0+%, TCI America™
SDP

Catalog No. A122825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A122825G 25 g
A1228250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A122825G Supplier TCI America Supplier No. A122825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6358-07-2
Molecular Formula C6H5ClN2O3
Molecular Weight (g/mol) 188.57
MDL Number MFCD00010300
InChI Key ZARYBZGMUVAJMK-UHFFFAOYSA-N
Synonym 5-Chloro-2-hydroxy-4-nitroaniline
PubChem CID 3894848
IUPAC Name 2-amino-4-chloro-5-nitrophenol
SMILES C1=C(C(=CC(=C1Cl)[N+](=O)[O-])O)N

Specifications

Melting Point 220°C
Quantity 25 g
Formula Weight 188.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-chloro-5-nitrophenol
RTECSNumber SJ5736000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.