Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-4-(ethylthio)butanoic acid | 67-21-0 | MFCD00063102 | 98.8% | 163.24 g·mol⁻¹ | C6H13NO2S | 5 G
SDP

Supplier:  Medchemexpress LLC HYW0172025G

Encompass_Preferred

2-Amino-4-(ethylthio)butanoic acid (CAS 67-21-0) is a methionine derivative used as an amino-acid analogue in biochemical research and synthetic applications. The compound is typically supplied as a white to off-white solid and is used in studies of methylation, protein synthesis, and metabolic pathway modulation.

  • Molecular formula C6H13NO2S.
  • Molar mass 163.24 g·mol⁻¹.
  • High purity (98.8%) as indicated by COA.
  • Supplied as a research reagent solid suitable for synthesis and assays.
  • Stable under recommended storage conditions for long-term use.

Catalog No. 50-003-00551


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.