Learn More
Medchemexpress LLC 2-amino-4-(ethylthio)butanoic acid | 67-21-0 | MFCD00063102 | 99.4% | 163.24 g·mol⁻1 | C6H13NO2S | 1 G
SDP
Catalog No. 5000300149
Encompass_Preferred
2-Amino-4-(ethylthio)butanoic acid is a methionine-derived amino acid analog provided as a high-purity research reagent for biochemical and pharmacological studies. It is offered as a stable powder with manufacturer-provided documentation to support purity, handling, and safety for laboratory use.
  • High purity (99.36%) suitable for analytical and preparative work.
  • Powder form with recommended storage at -20°C for long-term stability.
  • Solvent storage guidance: -80°C for 6 months, -20°C for 1 month.
  • Applicable as a methionine analog in metabolic and biochemical studies.
  • Supplied with COA and SDS for traceability and safety compliance.
Catalog No. 50-003-00149 Supplier Medchemexpress LLC Supplier No. HYW0172021G
2-Amino-4-(ethylthio)butanoic acid is a methionine-derived amino acid analog provided as a high-purity research reagent for biochemical and pharmacological studies. It is offered as a stable powder with manufacturer-provided documentation to support purity, handling, and safety for laboratory use.
  • High purity (99.36%) suitable for analytical and preparative work.
  • Powder form with recommended storage at -20°C for long-term stability.
  • Solvent storage guidance: -80°C for 6 months, -20°C for 1 month.
  • Applicable as a methionine analog in metabolic and biochemical studies.
  • Supplied with COA and SDS for traceability and safety compliance.