Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-4-(ethylthio)butanoic acid | 67-21-0 | MFCD00063102 | 99.4% | 163.24 g·mol⁻¹ | C6H13NO2S | 10 G
SDP

Catalog No. 5000299520
Encompass_Preferred

2-Amino-4-(ethylthio)butanoic acid is a methionine-derived amino acid analogue supplied as a research-grade biochemical reagent. It is intended for laboratory studies of amino acid metabolism, enzyme assays, and nutritional or pharmacological investigations; not for human or clinical use.

  • High purity (99.36%).
  • Molecular weight 163.24 g·mol⁻¹.
  • Soluble in water for aqueous assays.
  • Available in multiple pack sizes for laboratory experiments.
  • Provided with certificate of analysis and safety data sheet.

Catalog No. 50-002-99520 Supplier Medchemexpress LLC Supplier No. HYW01720210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-4-(ethylthio)butanoic acid is a methionine-derived amino acid analogue supplied as a research-grade biochemical reagent. It is intended for laboratory studies of amino acid metabolism, enzyme assays, and nutritional or pharmacological investigations; not for human or clinical use.

  • High purity (99.36%).
  • Molecular weight 163.24 g·mol⁻¹.
  • Soluble in water for aqueous assays.
  • Available in multiple pack sizes for laboratory experiments.
  • Provided with certificate of analysis and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.