Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-fluorobenzoic acid, 96%, Thermo Scientific™

Catalog No. AC293020050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC293020050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC293020050 Supplier Thermo Scientific Chemicals Supplier No. 293020050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-32-2
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
InChI Key LGPVTNAJFDUWLF-UHFFFAOYSA-N
Synonym 4-fluoroanthranilic acid, 2-amino-4-fluoro-benzoic acid, benzoic acid, 2-amino-4-fluoro, 2-amino-4-fluoro benzoic acid, 2-amino-4-fluorobenzoicacid, fa0, pubchem1323, amino-4-fluorobenzoic acid, acmc-209jz7, 2-amino-4fluorobenzoic acid
PubChem CID 2724967
IUPAC Name 2-amino-4-fluorobenzoic acid
SMILES C1=CC(=C(C=C1F)N)C(=O)O

Specifications

Melting Point 188.0°C to 196.0°C
Linear Formula H2NC6H3(F)COOH
Quantity 5 g
Formula Weight 155.13
Chemical Name or Material 2-Amino-4-fluorobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.