Learn More
Medchemexpress LLC 2-Amino-4-fluoropyridine | 944401-77-8 | 112.11 | 10 G
SDP
Catalog No. 5000446035
Encompass_Preferred
2-Amino-4-fluoropyridine (4-Fluoropyridin-2-amine) is a drug intermediate that can be used for the synthesis of reversible-covalent FGFR4 inhibitors.
  • Drug intermediate
  • Can be used for the synthesis of reversible-covalent FGFR4 inhibitors
Catalog No. 50-004-46035 Supplier Medchemexpress LLC Supplier No. HYW00511810G
2-Amino-4-fluoropyridine (4-Fluoropyridin-2-amine) is a drug intermediate that can be used for the synthesis of reversible-covalent FGFR4 inhibitors.
  • Drug intermediate
  • Can be used for the synthesis of reversible-covalent FGFR4 inhibitors