Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-methylbenzothiazole 98.0+%, TCI America™
SDP

Catalog No. A134425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A134425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A134425G Supplier TCI America Supplier No. A134425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1477-42-5
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00005793
InChI Key GRIATXVEXOFBGO-UHFFFAOYSA-N
Synonym 2-amino-4-methylbenzothiazole, 4-methylbenzo d thiazol-2-amine, 2-benzothiazolamine, 4-methyl, 4-methyl-2-aminobenzothiazole, 4-methyl-2-benzothiazolamine, benzothiazole, 2-amino-4-methyl, 4-methylbenzothiazol-2-ylamine, 2-amino-4-methyl benzothiazole, 4-methyl-2-aminobenzothiozole, unii-45v5f1b9z8
PubChem CID 15132
IUPAC Name 4-methyl-1,3-benzothiazol-2-amine
SMILES CC1=C2N=C(N)SC2=CC=C1

Specifications

Melting Point 137°C
Color White-Yellow
Quantity 25 g
Formula Weight 164.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-methylbenzothiazole
EINECSNumber (5)-5486
RTECSNumber DL2275000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.