Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-nitrophenol, 99%, Thermo Scientific™

Catalog No. AC400891000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC400891000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC400891000 Supplier Thermo Scientific Chemicals Supplier No. 400891000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-57-0
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00007695
InChI Key VLZVIIYRNMWPSN-UHFFFAOYSA-N
Synonym 2-hydroxy-5-nitroaniline, phenol, 2-amino-4-nitro, 4-nitro-2-aminophenol, p-nitro-o-aminophenol, 4-nitro-2-aminofenol, p-nitroaminofenol, 3-amino-4-hydroxynitrobenzene, 1-nitro-3-amino-4-hydroxybenzene, rodol 42, 2-amino-4-nitrofenol
PubChem CID 3613389
ChEBI CHEBI:82383
IUPAC Name 2-amino-4-nitrophenol
SMILES NC1=CC(=CC=C1O)[N+]([O-])=O

Specifications

Melting Point 143.0°C
Flash Point 100°C
Quantity 100 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: very soluble in ethyl alcohol, soluble in ether, methyl alcohol, hot benzene,, acetic acid
Formula Weight 154.12
Chemical Name or Material 2-Amino-4-nitrophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.