Learn More
Medchemexpress LLC 2-amino-4-phenylthiazole | 2010-06-2 | MFCD00039680 | 100.0% | 176.24 | C9H8N2S | 10 G
SDP
Catalog No. 5000299381
Encompass_Preferred
2-Amino-4-phenylthiazole is a thiazole-based synthetic intermediate used in medicinal chemistry and agrochemical research. It is an off-white to light-yellow solid with molecular formula C9H8N2S and molecular weight 176.24 g/mol. High-purity material is provided for reliable performance in small-molecule synthesis and heterocycle construction.
  • High purity suitable for research-grade synthesis.
  • Useful building block for heterocyclic compound synthesis.
  • Solid form simplifies handling and storage.
  • Compatible with standard organic synthetic procedures.
  • Well-characterized with CAS and MFCD identifiers for traceability.
Catalog No. 50-002-99381 Supplier Medchemexpress LLC Supplier No. HYW01668910G
2-Amino-4-phenylthiazole is a thiazole-based synthetic intermediate used in medicinal chemistry and agrochemical research. It is an off-white to light-yellow solid with molecular formula C9H8N2S and molecular weight 176.24 g/mol. High-purity material is provided for reliable performance in small-molecule synthesis and heterocycle construction.
  • High purity suitable for research-grade synthesis.
  • Useful building block for heterocyclic compound synthesis.
  • Solid form simplifies handling and storage.
  • Compatible with standard organic synthetic procedures.
  • Well-characterized with CAS and MFCD identifiers for traceability.