Learn More
Medchemexpress LLC 2-Amino-4-phenylthiazole | 2010-06-2 | MFCD00039680 | 100.0% | 176.24 g/mol | C9H8N2S | 1 G
SDP
Catalog No. 5000300797
Encompass_Preferred
2-Amino-4-phenylthiazole (CAS 2010-06-2) is a small-molecule synthetic intermediate used in medicinal chemistry for preparation of histone deacetylase (HDAC) inhibitor analogs and other pharmaceutical targets. Supplied as a characterized solid with supporting analytical documentation.
  • High purity (99.95%) verified by certificate of analysis.
  • Molecular weight 176.24 g/mol; formula C9H8N2S for stoichiometry and calculations.
  • Suitable as a synthetic intermediate for histone deacetylase (HDAC) inhibitor and SAHA analog synthesis.
  • Available in laboratory-scale pack sizes for research use.
  • Supplied with COA and SDS for quality and safety compliance.
Catalog No. 50-003-00797 Supplier Medchemexpress LLC Supplier No. HYW0166891G
2-Amino-4-phenylthiazole (CAS 2010-06-2) is a small-molecule synthetic intermediate used in medicinal chemistry for preparation of histone deacetylase (HDAC) inhibitor analogs and other pharmaceutical targets. Supplied as a characterized solid with supporting analytical documentation.
  • High purity (99.95%) verified by certificate of analysis.
  • Molecular weight 176.24 g/mol; formula C9H8N2S for stoichiometry and calculations.
  • Suitable as a synthetic intermediate for histone deacetylase (HDAC) inhibitor and SAHA analog synthesis.
  • Available in laboratory-scale pack sizes for research use.
  • Supplied with COA and SDS for quality and safety compliance.