Learn More
Medchemexpress LLC 2-amino-4-phenylthiazole | 2010-06-2 | MFCD00039680 | 100.0% | 176.24 g/mol | C9H8N2S | 5 G
SDP
Catalog No. 5000303778
Encompass_Preferred
2-Amino-4-phenylthiazole is an organic thiazole intermediate used in pharmaceutical and chemical synthesis. It is supplied as an off-white to light yellow solid with molecular formula C9H8N2S and molecular weight 176.24 g/mol. The manufacturer's certificate of analysis reports high purity by HPLC and confirms the identity by 1H NMR.
  • High reported purity suitable for synthetic applications.
  • 1H NMR consistent with stated structure.
  • Solid form simplifies handling and storage.
  • Suitable as a building block for small-molecule synthesis.
  • Provided with certificate of analysis for quality traceability.
Catalog No. 50-003-03778 Supplier Medchemexpress LLC Supplier No. HYW0166895G
2-Amino-4-phenylthiazole is an organic thiazole intermediate used in pharmaceutical and chemical synthesis. It is supplied as an off-white to light yellow solid with molecular formula C9H8N2S and molecular weight 176.24 g/mol. The manufacturer's certificate of analysis reports high purity by HPLC and confirms the identity by 1H NMR.
  • High reported purity suitable for synthetic applications.
  • 1H NMR consistent with stated structure.
  • Solid form simplifies handling and storage.
  • Suitable as a building block for small-molecule synthesis.
  • Provided with certificate of analysis for quality traceability.