Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-4-phosphonobutyric acid | 6323-99-5 | MFCD00013999 | 98.0% | 183.10 g/mol | C4H10NO5P | 50 MG
SDP

Catalog No. 5000212486
Encompass_Preferred

DL-AP4 (2-amino-4-phosphonobutyric acid) is a synthetic glutamate receptor antagonist used in central nervous system and visual system research. Supplied as a solid powder, it is intended for preparation of aqueous solutions and is commonly used in electrophysiology and pharmacology experiments to study glutamatergic signaling.

  • High chemical purity (98.0%).
  • Solid powder form for accurate weighing and long-term storage.
  • Soluble in water with ultrasonic warming to 60°C for preparation of stock solutions.
  • Molecular weight 183.10 g/mol for precise dose calculations.
  • CAS number 6323-99-5 for unambiguous substance identification.

Catalog No. 50-002-12486 Supplier Medchemexpress LLC Supplier No. HY10074350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-AP4 (2-amino-4-phosphonobutyric acid) is a synthetic glutamate receptor antagonist used in central nervous system and visual system research. Supplied as a solid powder, it is intended for preparation of aqueous solutions and is commonly used in electrophysiology and pharmacology experiments to study glutamatergic signaling.

  • High chemical purity (98.0%).
  • Solid powder form for accurate weighing and long-term storage.
  • Soluble in water with ultrasonic warming to 60°C for preparation of stock solutions.
  • Molecular weight 183.10 g/mol for precise dose calculations.
  • CAS number 6323-99-5 for unambiguous substance identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.