Learn More
Medchemexpress LLC 2-Amino-4-(trifluoromethyl)pyridine | 106447-97-6 | 99.99% | C6H5F3N2 | 5 G
SDP
Catalog No. 5000434278
Encompass_Preferred
2-Amino-4-(trifluoromethyl)pyridine is a biochemical reagent. It can be utilized as a biological material or an organic compound for research in life sciences.
  • Used in life science-related research
  • Available in solid form, off-white to yellow color
  • Soluble in DMSO (≥ 100 mg/mL)
Catalog No. 50-004-34278 Supplier Medchemexpress LLC Supplier No. HY755135G
2-Amino-4-(trifluoromethyl)pyridine is a biochemical reagent. It can be utilized as a biological material or an organic compound for research in life sciences.
  • Used in life science-related research
  • Available in solid form, off-white to yellow color
  • Soluble in DMSO (≥ 100 mg/mL)