Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-(4-nitrophenylsulfonyl)-thiazole, Thermo Scientific™

Catalog No. AC104221000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC104221000 100 g
AC104220250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104221000 Supplier Thermo Scientific Chemicals Supplier No. 104221000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39565-05-4
Molecular Formula C9H7N3O4S2
Molecular Weight (g/mol) 285.292
InChI Key GKTKCGAOXFHFTD-UHFFFAOYSA-N
Synonym 2-amino-5-4-nitrophenylsulfonyl thiazole, 2-amino-5-p-nitrophenylsulfonyl thiazole, 2-amino-5-p-nitrobenzenesulfonyl thiazole, 5-4-nitrophenyl sulfonyl-1,3-thiazol-2-amine, 2-thiazolamine, 5-4-nitrophenyl sulfonyl, thiazole, 2-amino-5-p-nitrophenyl sulfonyl, thiazole, 2-amino-5-p-nitrophenylsulfonyl, p-nitrophenyl-2-aminothiazol-5-ylsulphone, wln: t5n csj bz dswr dnw
PubChem CID 1535
IUPAC Name 5-(4-nitrophenyl)sulfonyl-1,3-thiazol-2-amine
SMILES C1=CC(=CC=C1[N+](=O)[O-])S(=O)(=O)C2=CN=C(S2)N

Specifications

Melting Point 222°C to 226°C
Color Yellow
Infrared Spectrum Authentic
Quantity 100 g
Formula Weight 285.28
Physical Form Powder
Chemical Name or Material 2-Amino-5-(4-nitrophenylsulfonyl)-thiazole
EINECSNumber 254-515-2
RTECSNumber XJ2870000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.