Learn More
Medchemexpress LLC 2-amino-5,6-dihydro-6-methyl-4H-1,3-thiazine hydrochloride | 21463-31-0 | MFCD00717539 | 99.0% | 166.67 g/mol | C5H11ClN2S | 1 MG
SDP
Catalog No. 5000211705
Encompass_Preferred
AMT hydrochloride is a research-grade selective inhibitor of inducible nitric oxide synthase (iNOS) with a reported Ki of approximately 4.2 nM. Supplied as a white to off-white solid, it is intended for laboratory research use; consult the safety data sheet for handling and storage recommendations (CAS 21463-31-0).
  • Selective iNOS inhibitor with reported Ki ≈ 4.2 nM.
  • High purity (~99.0%) suitable for research applications.
  • White to off-white solid powder for easy weighing and formulation.
  • Molecular weight 166.67 g/mol.
  • Stable when stored frozen; follow SDS storage conditions.
Catalog No. 50-002-11705 Supplier Medchemexpress LLC Supplier No. HY1012511MG
AMT hydrochloride is a research-grade selective inhibitor of inducible nitric oxide synthase (iNOS) with a reported Ki of approximately 4.2 nM. Supplied as a white to off-white solid, it is intended for laboratory research use; consult the safety data sheet for handling and storage recommendations (CAS 21463-31-0).
  • Selective iNOS inhibitor with reported Ki ≈ 4.2 nM.
  • High purity (~99.0%) suitable for research applications.
  • White to off-white solid powder for easy weighing and formulation.
  • Molecular weight 166.67 g/mol.
  • Stable when stored frozen; follow SDS storage conditions.