Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-(benzylthio)-1,3,4-thiadiazole 98.0+%, TCI America™
SDP

Catalog No. A26771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26771G 1 g
A26775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26771G Supplier TCI America Supplier No. A26771G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25660-71-3
Molecular Formula C9H9N3S2
Molecular Weight (g/mol) 223.312
MDL Number MFCD00184237
InChI Key BHIGBGKIAJJBGD-UHFFFAOYSA-N
Synonym 5-(Benzylthio)-1,3,4-thiadiazole-2-amine
PubChem CID 99285
IUPAC Name 5-benzylsulfanyl-1,3,4-thiadiazol-2-amine
SMILES C1=CC=C(C=C1)CSC2=NN=C(S2)N

Specifications

Melting Point 159°C
Color Yellow
Quantity 1 g
Formula Weight 223.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-(benzylthio)-1,3,4-thiadiazole
RTECSNumber XI2898500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.