Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-bromo-3-methoxypyrazine 99.0+%, TCI America™
SDP

Catalog No. A25621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A25621G Supplier TCI America Supplier No. A25621G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5900-13-0
Molecular Formula C5H6BrN3O
Molecular Weight (g/mol) 204.027
MDL Number MFCD08275726
InChI Key QDFJJHGAODFQMN-UHFFFAOYSA-N
PubChem CID 11344724
IUPAC Name 5-bromo-3-methoxypyrazin-2-amine
SMILES COC1=NC(=CN=C1N)Br

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 1 g
Formula Weight 204.03
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromo-3-methoxypyrazine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.