Learn More
Medchemexpress LLC 2-Amino-5-bromo-4-methylpyridine | 98198-48-2 | MFCD03427660 | 98% | 187.04 | 500 G
SDP
Catalog No. 5000386435
Encompass_Preferred
2-Amino-5-bromo-4-methylpyridine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also used as a pharmaceutical intermediate.
  • Used as a biochemical reagent
  • Can be used as a biological material or organic compound for life science related research
  • Used as a pharmaceutical intermediate
  • Targeted for biochemical assay reagents
  • Storage at 4°C, protected from light
  • In solvent: -80°C for 6 months; -20°C for 1 month (protected from light)
Catalog No. 50-003-86435 Supplier Medchemexpress LLC Supplier No. HY17562500G
2-Amino-5-bromo-4-methylpyridine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also used as a pharmaceutical intermediate.
  • Used as a biochemical reagent
  • Can be used as a biological material or organic compound for life science related research
  • Used as a pharmaceutical intermediate
  • Targeted for biochemical assay reagents
  • Storage at 4°C, protected from light
  • In solvent: -80°C for 6 months; -20°C for 1 month (protected from light)