Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-5-bromobenzophenone | 39859-36-4 | MFCD00059962 | 99.8% | 276.13 | C13H10BrNO | 25 G
SDP

Catalog No. 5000209072
Encompass_Preferred

2-amino-5-bromobenzophenone is a brominated benzophenone derivative used as an intermediate in organic and pharmaceutical synthesis. It is supplied as a high-purity solid suitable for medicinal chemistry and advanced organic synthesis applications.

  • Molecular formula C13H10BrNO.
  • Molecular weight 276.13.
  • Purity 99.8%.
  • Storage 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Pack sizes available 1 g, 5 g, 10 g, 25 g, 50 g, 100 g, and larger.

Catalog No. 50-002-09072 Supplier Medchemexpress LLC Supplier No. 25GHYW087426
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-amino-5-bromobenzophenone is a brominated benzophenone derivative used as an intermediate in organic and pharmaceutical synthesis. It is supplied as a high-purity solid suitable for medicinal chemistry and advanced organic synthesis applications.

  • Molecular formula C13H10BrNO.
  • Molecular weight 276.13.
  • Purity 99.8%.
  • Storage 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Pack sizes available 1 g, 5 g, 10 g, 25 g, 50 g, 100 g, and larger.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.