Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-5-bromobenzophenone | 39859-36-4 | MFCD00059962 | 99.8% | 276.13 g/mol | C13H10BrNO | 5 G
SDP

Catalog No. 5000208413
Encompass_Preferred

2-amino-5-bromobenzophenone is a brominated benzophenone derivative used as an intermediate in organic and pharmaceutical synthesis. It is commonly used in research for building chemical libraries and preparing substituted benzophenone derivatives.

  • Used as a synthesis intermediate in pharmaceutical chemistry.
  • High purity suitable for analytical and synthetic applications.
  • Available in multiple package sizes for small-scale research.
  • Chemical formula C13H10BrNO and molecular weight 276.13 g/mol.
  • CAS number 39859-36-4 for unambiguous identification.

Catalog No. 50-002-08413 Supplier Medchemexpress LLC Supplier No. HYW0874265G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-amino-5-bromobenzophenone is a brominated benzophenone derivative used as an intermediate in organic and pharmaceutical synthesis. It is commonly used in research for building chemical libraries and preparing substituted benzophenone derivatives.

  • Used as a synthesis intermediate in pharmaceutical chemistry.
  • High purity suitable for analytical and synthetic applications.
  • Available in multiple package sizes for small-scale research.
  • Chemical formula C13H10BrNO and molecular weight 276.13 g/mol.
  • CAS number 39859-36-4 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.