Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-amino-5-bromobenzophenone | 39859-36-4 | MFCD00059962 | 99.8% | 276.13 g/mol | C13H10BrNO | 50 G
SDP

Catalog No. 5000208412
Encompass_Preferred

2-Amino-5-bromobenzophenone is a brominated benzophenone derivative used as a synthetic intermediate in the preparation of pharmaceutical compounds and research chemistry.

  • Brominated benzophenone derivative.
  • High purity (99.82% by HPLC).
  • Cas number 39859-36-4.
  • Molecular formula C13H10BrNO; molecular weight 276.13 g/mol.
  • Available in multiple pack sizes, including 50 g.
  • Used as a building block for small-molecule synthesis.

Catalog No. 50-002-08412 Supplier Medchemexpress LLC Supplier No. HYW08742650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-5-bromobenzophenone is a brominated benzophenone derivative used as a synthetic intermediate in the preparation of pharmaceutical compounds and research chemistry.

  • Brominated benzophenone derivative.
  • High purity (99.82% by HPLC).
  • Cas number 39859-36-4.
  • Molecular formula C13H10BrNO; molecular weight 276.13 g/mol.
  • Available in multiple pack sizes, including 50 g.
  • Used as a building block for small-molecule synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.