Learn More
Medchemexpress LLC 2-Amino-5-bromothiazole hydrobromide | 61296-22-8 | 98.0% | C3H4Br2N2S | 100 G
SDP
Catalog No. 5000434209
Encompass_Preferred
2-Amino-5-bromothiazole hydrobromide is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent or as a stock solution at -80°C for 6 months or -20°C for 1 month.
  • Ships at room temperature in continental US; may vary elsewhere.
Catalog No. 50-004-34209 Supplier Medchemexpress LLC Supplier No. HY59343100G
2-Amino-5-bromothiazole hydrobromide is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent or as a stock solution at -80°C for 6 months or -20°C for 1 month.
  • Ships at room temperature in continental US; may vary elsewhere.