Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-bromothiazole monohydrobromide, 95%

Catalog No. AC156000250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156000250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC156000250 Supplier Thermo Scientific Chemicals Supplier No. 156000250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 61296-22-8 | C3H3BrN2S·HBr | 259.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61296-22-8
Molecular Formula C3H3BrN2S·HBr
Molecular Weight (g/mol) 259.95
MDL Number MFCD00012712
InChI Key NUSVDASTCPBUIP-UHFFFAOYSA-N
Synonym 2-amino-5-bromothiazole hydrobromide, 2-amino-5-bromothiazole monohydrobromide, 5-bromothiazol-2-amine hydrobromide, 5-bromo-1,3-thiazol-2-amine hydrobromide, 5-bromo-thiazol-2-ylamine hydrobromide, 2-amino-5-bromothiazole hbr, 2-amino-5-bromo-1,3-thiazole hydrobromide, 2-thiazolamine, 5-bromo-, monohydrobromide, 5-bromo-1,3-thiazole-2-ylamine, bromide, acmc-209mr7
PubChem CID 2723848
IUPAC Name 5-bromo-1,3-thiazol-2-amine;hydrobromide
SMILES C1=C(SC(=N1)N)Br.Br

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 165°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 25 g
Formula Weight 259.95
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromothiazole monohydrobromide, 95%
Show More Show Less
EINECSNumber 262-699-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.