Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Amino-5-chloro-2'-fluorobenzophenone | 784-38-3 | 249.67 | 50 G
SDP

Catalog No. 5000448869
Encompass_Preferred

2-Amino-5-chloro-2'-fluorobenzophenone is a chemical precursor used in the synthesis of Midazolam. This product is intended for research use only.

  • Precursor in Midazolam synthesis
  • Solid appearance
  • Light yellow to yellow color
  • Purity of 99.97%
  • Molecular weight: 249.67
  • CAS number: 784-38-3
  • Molecular formula: C13H9ClFNO
  • Melting point: 95-98°C
  • Boiling point: 207°C at 760 mmHg

Catalog No. 50-004-48869 Supplier Medchemexpress LLC Supplier No. HYW00217250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Amino-5-chloro-2'-fluorobenzophenone is a chemical precursor used in the synthesis of Midazolam. This product is intended for research use only.

  • Precursor in Midazolam synthesis
  • Solid appearance
  • Light yellow to yellow color
  • Purity of 99.97%
  • Molecular weight: 249.67
  • CAS number: 784-38-3
  • Molecular formula: C13H9ClFNO
  • Melting point: 95-98°C
  • Boiling point: 207°C at 760 mmHg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.