Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-chloro-4-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. A29455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29455G 5 g
A29451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29455G Supplier TCI America Supplier No. A29455G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 36936-27-3 | C6H7ClN2 | 142.59 g/mol

Chemical Identifiers

CAS 36936-27-3
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD06410759
InChI Key DGRCJSWXMGENMG-UHFFFAOYSA-N
Synonym 2-Amino-5-chloro-4-picoline
PubChem CID 318186
IUPAC Name 5-chloro-4-methylpyridin-2-amine
SMILES CC1=CC(N)=NC=C1Cl

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 5 g
Formula Weight 142.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-chloro-4-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.