Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-chloro-6-methylpyridine, 97%, Thermo Scientific Chemicals

Catalog No. AAH5850106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5850106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5850106 Supplier Thermo Scientific Chemicals Supplier No. H5850106
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 36936-23-9 | Molecular Formula: C6H7ClN2 | Molecular Weight: 142.586 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36936-23-9
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.586
MDL Number MFCD09453335
InChI Key SHIKRPPKGCWKJO-UHFFFAOYSA-N
Synonym 2-amino-5-chloro-6-methylpyridine, 2-amino-5-chloro-6-picoline, 5-chloro-6-methyl-2-pyridinamine, 6-amino-3-chloropicoline, 2-amino-5-chloro-6-methyl pyridine, 5-chloro-6-methyl-pyridin-2-ylamine, 2-pyridinamine, 5-chloro-6-methyl, pubchem5718, 6-amino-3-chloro-2-picoline, 5-chloro-6-methyl-pyridineamine
PubChem CID 12251007
IUPAC Name 5-chloro-6-methylpyridin-2-amine
SMILES CC1=C(C=CC(=N1)N)Cl

Specifications

Melting Point 70°C to 75°C
Quantity 5 g
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 2-Amino-5-chloro-6-methylpyridine
Hazard Category H302-H318
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.