Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Amino-5-chlorobenzophenone | 719-59-5 | 99.98% | C13H10ClNO | 500 G
SDP

Catalog No. 5000433397
Encompass_Preferred

2-Amino-5-chlorobenzophenone is a substituted benzophenone and can be used in the synthesis of benzodiazepines. The derivative has skeletal muscle relaxation activity. It is a solid that appears light yellow to yellow in color.

  • Used in the synthesis of benzodiazepines
  • Derivative exhibits skeletal muscle relaxation activity
  • Solid, light yellow to yellow appearance
  • High purity of 99.98%
  • Molecular weight of 231.68
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-004-33397 Supplier Medchemexpress LLC Supplier No. HY77785500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-5-chlorobenzophenone is a substituted benzophenone and can be used in the synthesis of benzodiazepines. The derivative has skeletal muscle relaxation activity. It is a solid that appears light yellow to yellow in color.

  • Used in the synthesis of benzodiazepines
  • Derivative exhibits skeletal muscle relaxation activity
  • Solid, light yellow to yellow appearance
  • High purity of 99.98%
  • Molecular weight of 231.68
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.