Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-amino-5-chloropyrazine

Catalog No. CDS005437 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS005437 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS005437 Supplier Sigma Aldrich Supplier No. CDS005437100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Hydrogen-bonded complexes of pentachlorocyclopropane with the bases acetonitrile, ammonia, monomethylamine and dimethylamine have been isolated.pentachlorocyclopropane has been used in the preparation of:
  • Tetrachlorocyclopropene via reaction with 18m aqueous koh at 80-85c
  • Substituted phenyltrichlorocyclopropene derivatives

Specifications

CAS 33332-29-5
Molecular Formula C4H4ClN3
Linear Formula C4H4ClN3
MDL Number MFCD03423596
Quantity 5 g
Molecular Weight (g/mol) 129.55
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.