Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-(diethylamino)toluene Monohydrochloride 98.0+%, TCI America™
SDP

Catalog No. A028925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A028925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A028925G Supplier TCI America Supplier No. A028925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory reagent used for research applications.

Specifications

CAS 2051-79-8
Color White
Molecular Formula C11H19ClN2
Quantity 25 g
MDL Number MFCD00035466
UN Number 2811
Synonym 2-amino-5-diethylamino toluene monohydrochloride, 4-n,n-diethyl-2-methyl-p-phenylenediamine monohydrochloride, 1,4-benzenediamine, n4,n4-diethyl-2-methyl-, monohydrochloride, unii-39mih70e0i, n4,n4-diethyl-2-methylbenzene-1,4-diamine hydrochloride, 2-amino-5-diethylaminotoluene monohydrochloride, 4-amino-3-methylphenyl diethylamine, chloride, 4-diethylamino-o-toluidine hydrochloride, n,n-diethyltoluene-2,5-diamine hydrochloride, 1,4-benzenediamine, n4,n4-diethyl-2-methyl-, hydrochloride 1:1
InChI Key MPLZNPZPPXERDA-UHFFFAOYSA-N
SMILES CCN(CC)C1=CC(=C(C=C1)N)C.Cl
IUPAC Name 4-N,4-N-diethyl-2-methylbenzene-1,4-diamine;hydrochloride
Molecular Weight (g/mol) 214.737
PubChem CID 108623
Formula Weight 214.74
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-(diethylamino)toluene Monohydrochloride
Show More Show Less
EINECSNumber (3)-0279
RTECSNumber SS9560000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.