Learn More
Accela Chembio Inc 2-amino-5-fluorobenzoic Acid | 100g | 446-08-2 | MFCD00055566 | 97+% | Shelf Life: 1260 Days | Light Sensitive/+4
Supplier:  Accela Chembio Inc SY004954100G
Encompass_Preferred
2-amino-5-fluorobenzoic Acid | 100g | 446-08-2 | MFCD00055566 | 97+% | Shelf Life: 1260 Days | Light Sensitive/+4
Catalog No. 50-354-177