Learn More
Medchemexpress LLC 2-Amino-5-fluorobenzoic acid | 446-08-2 | 99.98% | 155.13 | 25 G
SDP
Catalog No. 5000402540
Encompass_Preferred
2-Amino-5-fluorobenzoic acid is a biochemical reagent used as a biological material or organic compound for life science related research. It is a toxic antitimetabolite for the tryptophan pathway in yeast, capable of counterselecting for TRP1, a genetic marker in *S. cerevisiae*. TRP1-lacking strains are resistant to its feedback inhibition, allowing for growth-based positive selection of the TRP1 marker. It is frequently employed in genetic procedures involving plasma manip calculations.
  • Can be used as a biological material or organic compound
  • Toxic antitimetabolite for the tryptophan pathway in yeast
  • Used to counterselect for TRP1, a common genetic marker in *S. cerevisiae*
  • Frequently used in genetic procedures involving plasma manipulations
Catalog No. 50-004-02540 Supplier Medchemexpress LLC Supplier No. 25GHY34053
2-Amino-5-fluorobenzoic acid is a biochemical reagent used as a biological material or organic compound for life science related research. It is a toxic antitimetabolite for the tryptophan pathway in yeast, capable of counterselecting for TRP1, a genetic marker in *S. cerevisiae*. TRP1-lacking strains are resistant to its feedback inhibition, allowing for growth-based positive selection of the TRP1 marker. It is frequently employed in genetic procedures involving plasma manip calculations.
  • Can be used as a biological material or organic compound
  • Toxic antitimetabolite for the tryptophan pathway in yeast
  • Used to counterselect for TRP1, a common genetic marker in *S. cerevisiae*
  • Frequently used in genetic procedures involving plasma manipulations