Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Amino-5-fluorophenol | 53981-24-1 | 127.12 | 100 G
SDP

Catalog No. 5000460337
Encompass_Preferred

2-Amino-5-fluorophenol is an isomer of aminofluorophenol and a HONO generator. It reacts with nitrogen dioxide (NO2) via two pathways-hydrogen abstraction from the hydroxyl group and hydrogen abstraction from the amino group-to form nitrous acid (HONO). Both the amino and hydroxyl groups participate in the reaction in the transition state.

  • Isomer of aminofluorophenol
  • HONO generator
  • Reacts with nitrogen dioxide to form nitrous acid
  • Appears as a solid
  • Color is light brown to gray
  • Molecular formula: C6H6FNO

Catalog No. 50-004-60337 Supplier Medchemexpress LLC Supplier No. HYW007345100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Amino-5-fluorophenol is an isomer of aminofluorophenol and a HONO generator. It reacts with nitrogen dioxide (NO2) via two pathways-hydrogen abstraction from the hydroxyl group and hydrogen abstraction from the amino group-to form nitrous acid (HONO). Both the amino and hydroxyl groups participate in the reaction in the transition state.

  • Isomer of aminofluorophenol
  • HONO generator
  • Reacts with nitrogen dioxide to form nitrous acid
  • Appears as a solid
  • Color is light brown to gray
  • Molecular formula: C6H6FNO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.