Learn More
Medchemexpress LLC 2-Amino-5-fluorophenol | 53981-24-1 | 127.12 | 5 G
SDP
Catalog No. 5000460278
Encompass_Preferred
2-Amino-5-fluorophenol is an isomer of aminofluorophenol and also a HONO generator. It reacts with nitrogen dioxide (NO2) via two pathways to form nitrous acid (HONO).
  • Hydrogen abstraction from the hydroxyl group and hydrogen abstraction from the amino group.
  • Both the amino and hydroxyl groups participate in the reaction in the transition state.
Catalog No. 50-004-60278 Supplier Medchemexpress LLC Supplier No. HYW0073455G
2-Amino-5-fluorophenol is an isomer of aminofluorophenol and also a HONO generator. It reacts with nitrogen dioxide (NO2) via two pathways to form nitrous acid (HONO).
  • Hydrogen abstraction from the hydroxyl group and hydrogen abstraction from the amino group.
  • Both the amino and hydroxyl groups participate in the reaction in the transition state.